accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 54557-81-2

IUPAC Name: 4-(2,3-dihydro-1,4-benzodioxin-6-yl)-4-oxobutanoate
Synonyms: 4-(2,3-dihydro-1,4-benzodioxin-6-yl)-4-oxobutanoate
Molecular Weight (g/mol): 235.22
Molecular Formula: C12H11O5
InChi Key: LMDXEMFSAHAGGP-UHFFFAOYSA-M
SMILES: [O-]C(=O)CCC(=O)C1=CC=C2OCCOC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results