CAS RN 54571-67-4
IUPAC Name:
sodium (2S)-5-oxopyrrolidine-2-carboxylate
Synonyms:
sodium pca
Molecular Weight (g/mol):
151.1
Molecular Formula:
C5H6NNaO3
InChi Key:
CRPCXAMJWCDHFM-DFWYDOINSA-M
SMILES:
[Na+].[O-]C(=O)[C@@H]1CCC(=O)N1
1
–
1
of
1
results
1
Thermo Scientific Chemicals 2-Pyrrolidone-5-carboxylic acid, sodium salt, 50 wt% solution in water
CAS: 54571-67-4 | C5H6NNaO3 | 151.10 g/mol