CAS RN 54589-54-7
IUPAC Name:
ethyl 3-(chloromethyl)benzoate
Synonyms:
ethyl 3-(chloromethyl)benzoate
Molecular Weight (g/mol):
198.65
Molecular Formula:
C10H11ClO2
InChi Key:
ZJNVMXXFCNKXLT-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=CC(CCl)=CC=C1
1
–
1
of
1
results
1
Ethyl 3-(Chloromethyl)benzoate 96.0+%, TCI America™
CAS: 54589-54-7 Molecular Formula: C10H11ClO2 Molecular Weight (g/mol): 198.646 InChI Key: ZJNVMXXFCNKXLT-UHFFFAOYSA-N Synonym: 3-(Chloromethyl)benzoic Acid Ethyl Ester PubChem CID: 14812190 IUPAC Name: ethyl 3-(chloromethyl)benzoate SMILES: CCOC(=O)C1=CC(=CC=C1)CCl