accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3CH3H3C

CAS RN 5460-08-2

IUPAC Name: ethyl 2-(2,4,6-trimethylphenyl)acetate
Synonyms: ethyl 2-(2,4,6-trimethylphenyl)acetate
Molecular Weight (g/mol): 206.29
Molecular Formula: C13H18O2
InChi Key: OFZZPFNYAMIBNU-UHFFFAOYSA-N
SMILES: CCOC(=O)CC1=C(C)C=C(C)C=C1C
Boiling Point 
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results