accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONN

CAS RN 54605-72-0

IUPAC Name: 1-phenyl-1H-pyrazole-4-carbaldehyde
Synonyms: 1-phenylpyrazole-4-carbaldehyde
Molecular Weight (g/mol): 172.19
Molecular Formula: C10H8N2O
InChi Key: PHVRLPFVPVKYOI-UHFFFAOYSA-N
SMILES: O=CC1=CN(N=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results