CAS RN 54613-99-9
IUPAC Name:
2-{[(tert-butoxy)carbonyl]amino}-6-({[(2-chlorophenyl)methoxy]carbonyl}amino)hexanoic acid
Synonyms:
2-[(tert-butoxycarbonyl)amino]-6-({[(2-chlorophenyl)methoxy]carbonyl}amino)hexanoic acid
Molecular Weight (g/mol):
414.88
Molecular Formula:
C19H27ClN2O6
InChi Key:
ATUMDPHEFWGCJF-UHFFFAOYNA-N
SMILES:
CC(C)(C)OC(=O)NC(CCCCNC(=O)OCC1=CC=CC=C1Cl)C(O)=O
1
–
1
of
1
results
1
Boc-Lys 2-Cl-Z -OH, Advanced ChemTech
CAS: 54613-99-9 Molecular Formula: C19H27ClN2O6 Molecular Weight (g/mol): 414.88 InChI Key: ATUMDPHEFWGCJF-UHFFFAOYNA-N IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-6-({[(2-chlorophenyl)methoxy]carbonyl}amino)hexanoic acid SMILES: CC(C)(C)OC(=O)NC(CCCCNC(=O)OCC1=CC=CC=C1Cl)C(O)=O