accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHNClClNOO

CAS RN 5462-30-6

IUPAC Name: N-(4,5-dichloro-2-nitrophenyl)acetamide
Synonyms: N-(4,5-dichloro-2-nitrophenyl)acetamide
Molecular Weight (g/mol): 249.05
Molecular Formula: C8H6Cl2N2O3
InChi Key: ZEGRPTYRAGSSBH-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC(Cl)=C(Cl)C=C1[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results