CAS RN 54623-25-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Promotions Available
2,3,5-Tri-O-benzyl-D-ribofuranose, 98%
2,3,5-Tri-O-benzyl-D-ribofuranose, 98%
CAS: 54623-25-5 Molecular Formula: C26H28O5 MDL Number: MFCD03425640