accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H2NNH2H3C

CAS RN 54628-21-6

IUPAC Name: 4-[2-(4-amino-2-methylphenyl)ethyl]-3-methylaniline
Synonyms: 4-[2-(4-amino-2-methylphenyl)ethyl]-3-methylaniline
Molecular Weight (g/mol): 240.35
Molecular Formula: C16H20N2
InChi Key: ISESBQNCWCFFFR-UHFFFAOYSA-N
SMILES: CC1=C(CCC2=C(C)C=C(N)C=C2)C=CC(N)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results