accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 54629-13-9

IUPAC Name: 2-(4-fluorophenoxy)pyridine-3-carboxylate
Synonyms: 2-(4-fluorophenoxy)pyridine-3-carboxylate
Molecular Weight (g/mol): 232.19
Molecular Formula: C12H7FNO3
InChi Key: SDZUYDOXBXHDCE-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=CN=C1OC1=CC=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results