accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClCH3ONHN

CAS RN 5464-78-8

IUPAC Name: 1-(2-methoxyphenyl)piperazine hydrochloridyl
Synonyms: 1-(2-methoxyphenyl)piperazine hydrochloridyl
Molecular Weight (g/mol): 227.71
Molecular Formula: C11H16ClN2O
InChi Key: HOVMRUUGNVCBPH-UHFFFAOYSA-N
SMILES: [Cl].COC1=CC=CC=C1N1CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results