accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 54648-79-2

IUPAC Name: N,N'-bis(propan-2-yl)methoxymethanimidamide
Synonyms: N,N'-diisopropylmethoxymethanimidamide
Molecular Weight (g/mol): 158.25
Molecular Formula: C8H18N2O
InChi Key: PUVRRPLSJKDMKH-UHFFFAOYSA-N
SMILES: COC(NC(C)C)=NC(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results