accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5465-96-3

IUPAC Name: N-nitro-4,5-dihydro-1H-imidazol-2-amine
Synonyms: N-nitro-4,5-dihydro-1H-imidazol-2-amine
Molecular Weight (g/mol): 130.11
Molecular Formula: C3H6N4O2
InChi Key: DJZWNSRUEJSEEB-UHFFFAOYSA-N
SMILES: [O-][N+](=O)NC1=NCCN1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results