accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 54655-07-1

IUPAC Name: lithium(1+) (trimethylsilyl)ethyne
Synonyms: lithium(1+) (trimethylsilyl)ethyne
Molecular Weight (g/mol): 104.15
Molecular Formula: C5H9LiSi
InChi Key: WDDOQHLJFOUQMW-UHFFFAOYSA-N
SMILES: [Li+].C[Si](C)(C)C#[C-]
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)