accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH3NNH3NOSOOO

CAS RN 5466-22-8

IUPAC Name: bis(cyanomethanaminium) sulfate
Synonyms: bis(cyanomethanaminium) sulfate
Molecular Weight (g/mol): 210.21
Molecular Formula: C4H10N4O4S
InChi Key: NQHWLBZZBSYIPI-UHFFFAOYSA-N
SMILES: [NH3+]CC#N.[NH3+]CC#N.[O-]S([O-])(=O)=O
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (2)
  • (3)
  • (2)
Grade 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)