accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 54660-00-3

IUPAC Name: 3,6-diphenyl-1H,2H,4H,5H-pyrrolo[3,4-c]pyrrole-1,4-dione
Synonyms: 3,6-diphenyl-2H,5H-pyrrolo[3,4-c]pyrrole-1,4-dione
Molecular Weight (g/mol): 288.31
Molecular Formula: C18H12N2O2
InChi Key: UWHBZLMSPZBZHY-UHFFFAOYSA-N
SMILES: O=C1NC(=C2C(=O)NC(=C12)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results