accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SnH3CCH3S

CAS RN 54663-78-4

IUPAC Name: tributyl(thiophen-2-yl)stannane
Synonyms: tributyl(thiophen-2-yl)stannane
Molecular Weight (g/mol): 373.19
Molecular Formula: C16H30SSn
InChi Key: UKTDFYOZPFNQOQ-UHFFFAOYSA-N
SMILES: CCCC[Sn](CCCC)(CCCC)C1=CC=CS1
Boiling Point 
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)