accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HNHCl

CAS RN 54677-53-1

IUPAC Name: 2-methylpyrrolidine hydrochloride
Synonyms: 2-methylpyrrolidine hydrochloride
Molecular Weight (g/mol): 121.61
Molecular Formula: C5H12ClN
InChi Key: JNEIFWYJFOEKIM-UHFFFAOYNA-N
SMILES: Cl.CC1CCCN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)