accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5468-37-1

IUPAC Name: 2-amino-1-phenylethan-1-one hydrochloride
Synonyms: phenacylamine hydrochloride
Molecular Weight (g/mol): 171.62
Molecular Formula: C8H10ClNO
InChi Key: CVXGFPPAIUELDV-UHFFFAOYSA-N
SMILES: Cl.NCC(=O)C1=CC=CC=C1
Percent Purity 
  • (1)
  • (2)
  • (1)
Quantity 
  • (3)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results