accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONONCl

CAS RN 5470-18-8

IUPAC Name: 2-chloro-3-nitropyridine
Synonyms: 2-chloro-3-nitropyridine
Molecular Weight (g/mol): 158.54
Molecular Formula: C5H3ClN2O2
InChi Key: UUOLETYDNTVQDY-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CN=C1Cl
Molecular Weight (g/mol) 
  • (3)
  • (5)
Percent Purity 
  • (2)
  • (4)
  • (2)
Quantity 
  • (2)
  • (3)
  • (1)
  • (2)
Physical Form 
  • (4)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (5)
  • (3)
  • (5)
  • (2)
  • (4)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (4)
  • (1)