accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 54711-38-5

IUPAC Name: 7-(diethylamino)-4-(hydroxymethyl)-2H-chromen-2-one
Synonyms: 7-(diethylamino)-4-(hydroxymethyl)chromen-2-one
Molecular Weight (g/mol): 247.29
Molecular Formula: C14H17NO3
InChi Key: NMZSXNOCNJMJQT-UHFFFAOYSA-N
SMILES: CCN(CC)C1=CC=C2C(CO)=CC(=O)OC2=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results