accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 54714-11-3

IUPAC Name: diphenyl-2H-5λ⁶-thieno[3,4-d][1,3]dioxole-2,5,5-trione
Synonyms: diphenyl-5λ⁶-thieno[3,4-d][1,3]dioxole-2,5,5-trione
Molecular Weight (g/mol): 326.32
Molecular Formula: C17H10O5S
InChi Key: UEOYFCIGZWQXTP-UHFFFAOYSA-N
SMILES: O=C1OC2=C(C3=CC=CC=C3)S(=O)(=O)C(=C2O1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results