accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5472-49-1

IUPAC Name: hydrogen 1-(3-chloropropyl)piperidine chloride
Synonyms: hydrogen 1-(3-chloropropyl)piperidine chloride
Molecular Weight (g/mol): 198.13
Molecular Formula: C8H17Cl2N
InChi Key: OBOBUDMMFXRNDO-UHFFFAOYSA-N
SMILES: [H+].[Cl-].ClCCCN1CCCCC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)