accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 54760-75-7

IUPAC Name: 4-[(1E)-2-(4-aminophenyl)ethenyl]aniline
Synonyms: 4-[(1E)-2-(4-aminophenyl)ethenyl]aniline
Molecular Weight (g/mol): 210.28
Molecular Formula: C14H14N2
InChi Key: KOGDFDWINXIWHI-OWOJBTEDSA-N
SMILES: NC1=CC=C(\C=C\C2=CC=C(N)C=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results