accessibility menu, dialog, popup

UserName

Viewing profile as user:
O

CAS RN 548-39-0

IUPAC Name: 1H-phenalen-1-one
Synonyms: perinaphthenone
Molecular Weight (g/mol): 180.21
Molecular Formula: C13H8O
InChi Key: WWBGWPHHLRSTFI-UHFFFAOYSA-N
SMILES: O=C1C=CC2=C3C(C=CC=C13)=CC=C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results