accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2H2NNH2

CAS RN 548-61-8

IUPAC Name: 4-[bis(4-aminophenyl)methyl]aniline
Synonyms: 4-[bis(4-aminophenyl)methyl]aniline
Molecular Weight (g/mol): 289.38
Molecular Formula: C19H19N3
InChi Key: ADUMIBSPEHFSLA-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1)C(C1=CC=C(N)C=C1)C1=CC=C(N)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results