accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NOOH2N

CAS RN 54802-10-7

IUPAC Name: 3-amino-1-benzofuran-2-carboxamide
Synonyms: 3-amino-1-benzofuran-2-carboxamide
Molecular Weight (g/mol): 176.18
Molecular Formula: C9H8N2O2
InChi Key: FXOYSSLDDDZIJN-UHFFFAOYSA-N
SMILES: NC(=O)C1=C(N)C2=CC=CC=C2O1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results