accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 54802-12-9

IUPAC Name: 2-(2-cyanophenoxy)acetamide
Synonyms: 2-(2-cyanophenoxy)acetamide
Molecular Weight (g/mol): 176.18
Molecular Formula: C9H8N2O2
InChi Key: RYOYNMJXEHNYAG-UHFFFAOYSA-N
SMILES: NC(=O)COC1=CC=CC=C1C#N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results