accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOOHOHOHO

CAS RN 5488-16-4

IUPAC Name: 2-[4-(carboxymethyl)-2,5-dihydroxyphenyl]acetic acid
Synonyms: [4-(carboxymethyl)-2,5-dihydroxyphenyl]acetic acid
Molecular Weight (g/mol): 226.18
Molecular Formula: C10H10O6
InChi Key: MCLKERLHVBEZIW-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC(O)=C(CC(O)=O)C=C1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results