accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5490-27-7

IUPAC Name: bis(N''-[(1R,2R,3S,4R,5R,6S)-3-[(diaminomethylidene)amino]-4-{[(2R,3R,4R,5S)-3-{[(2S,3S,4S,5R,6S)-4,5-dihydroxy-6-(hydroxymethyl)-3-(methylamino)oxan-2-yl]oxy}-4-hydroxy-4-(hydroxymethyl)-5-methyloxolan-2-yl]oxy}-2,5,6-trihydroxycyclohexyl]guanidine); tris(sulfuric acid)
Synonyms: bis(N''-[(1R,2R,3S,4R,5R,6S)-3-[(diaminomethylidene)amino]-4-{[(2R,3R,4R,5S)-3-{[(2S,3S,4S,5R,6S)-4,5-dihydroxy-6-(hydroxymethyl)-3-(methylamino)oxan-2-yl]oxy}-4-hydroxy-4-(hydroxymethyl)-5-methyloxolan-2-yl]oxy}-2,5,6-trihydroxycyclohexyl]guanidine); tris(sulfuric acid)
Molecular Weight (g/mol): 1461.41
Molecular Formula: C42H88N14O36S3
InChi Key: CZWJCQXZZJHHRH-YCRXJPFRSA-N
SMILES: OS(O)(=O)=O.OS(O)(=O)=O.OS(O)(=O)=O.CN[C@@H]1[C@H](O[C@H]2[C@H](O[C@H]3[C@H](O)[C@@H](O)[C@H](N=C(N)N)[C@@H](O)[C@@H]3N=C(N)N)O[C@@H](C)[C@]2(O)CO)O[C@@H](CO)[C@H](O)[C@H]1O.CN[C@@H]1[C@H](O[C@H]2[C@H](O[C@H]3[C@H](O)[C@@H](O)[C@H](N=C(N)N)[C@@H](O)[C@@H]3N=C(N)N)O[C@@H](C)[C@]2(O)CO)O[C@@H](CO)[C@H](O)[C@H]1O
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)