accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3BrOHN

CAS RN 54923-31-8

IUPAC Name: 5-bromo-6-methyl-1,2-dihydropyridin-2-one
Synonyms: 5-bromo-6-methyl-1H-pyridin-2-one
Molecular Weight (g/mol): 188.02
Molecular Formula: C6H6BrNO
InChi Key: UJHCRBDEJPQFIA-UHFFFAOYSA-N
SMILES: CC1=C(Br)C=CC(=O)N1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results