CAS RN 549476-61-1
IUPAC Name:
N-cyclopropyl-4-nitrobenzene-1-sulfonamide
Synonyms:
N-cyclopropyl-4-nitrobenzenesulfonamide
Molecular Weight (g/mol):
242.25
Molecular Formula:
C9H10N2O4S
InChi Key:
SFXOKDPLGLHGIX-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=CC=C(C=C1)S(=O)(=O)NC1CC1
1
–
1
of
1
results
1
Promotions Available
N-Cyclopropyl-4-nitrobenzenesulfonamide, 97%, Thermo Scientific™
N-Cyclopropyl-4-nitrobenzenesulfonamide, 97%, Thermo Scientific™
CAS: 549476-61-1 Molecular Formula: C9H10N2O4S Molecular Weight (g/mol): 242.249 MDL Number: MFCD00458256 InChI Key: SFXOKDPLGLHGIX-UHFFFAOYSA-N PubChem CID: 3434716 IUPAC Name: N-cyclopropyl-4-nitrobenzenesulfonamide SMILES: C1CC1NS(=O)(=O)C2=CC=C(C=C2)[N+](=O)[O-]