accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 549544-24-3

IUPAC Name: N-(3-bromophenyl)-4-chlorobutanamide
Synonyms: N-(3-bromophenyl)-4-chlorobutanamide
Molecular Weight (g/mol): 276.56
Molecular Formula: C10H11BrClNO
InChi Key: GHKCNIGXORDORM-UHFFFAOYSA-N
SMILES: ClCCCC(=O)NC1=CC(Br)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results