accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2BrFFF

CAS RN 54962-75-3

IUPAC Name: 3-bromo-5-(trifluoromethyl)aniline
Synonyms: 3-bromo-5-(trifluoromethyl)aniline
Molecular Weight (g/mol): 240.02
Molecular Formula: C7H5BrF3N
InChi Key: HJTLKVYOWNTDPF-UHFFFAOYSA-N
SMILES: NC1=CC(Br)=CC(=C1)C(F)(F)F
Boiling Point 
  • (1)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (4)