accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 55-10-7

IUPAC Name: (2S)-2-hydroxy-2-(4-hydroxy-3-methoxyphenyl)acetate
Synonyms: vanillylmandelic acid
Molecular Weight (g/mol): 197.17
Molecular Formula: C9H9O5
InChi Key: CGQCWMIAEPEHNQ-QMMMGPOBSA-M
SMILES: COC1=CC(=CC=C1O)[C@H](O)C([O-])=O
Molecular Weight (g/mol) 
  • (5)
  • (3)
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (3)
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (5)
  • (3)
  • (2)
  • (3)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)