accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3NNH3C(E/Z)

CAS RN 55-80-1

IUPAC Name: N,N-dimethyl-4-[2-(3-methylphenyl)diazen-1-yl]aniline
Synonyms: N,N-dimethyl-4-[2-(3-methylphenyl)diazen-1-yl]aniline
Molecular Weight (g/mol): 239.32
Molecular Formula: C15H17N3
InChi Key: LVTFSVIRYMXRSR-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=C(C=C1)N=NC1=CC=CC(C)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results