accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 55-81-2

IUPAC Name: 2-(4-methoxyphenyl)ethan-1-amine
Synonyms: 4-methoxyphenethylamine homoanisylamine 4-methoxy-2-phenethylamine 4-methoxy-β-phenylethylamine O-methyltyramine
Molecular Weight (g/mol): 151.21
Molecular Formula: C9H13NO
InChi Key: LTPVSOCPYWDIFU-UHFFFAOYSA-N
SMILES: COC1=CC=C(CCN)C=C1
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)