accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 55-97-0

IUPAC Name: trimethyl[6-(trimethylazaniumyl)hexyl]azanium dibromide
Synonyms: hexamethonium bromide [ban:inn:jan]
Molecular Weight (g/mol): 362.19
Molecular Formula: C12H30Br2N2
InChi Key: FAPSXSAPXXJTOU-UHFFFAOYSA-L
SMILES: [Br-].[Br-].C[N+](C)(C)CCCCCC[N+](C)(C)C
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)