accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNOO

CAS RN 550-44-7

IUPAC Name: 2-methyl-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms: 2-methylisoindole-1,3-dione
Molecular Weight (g/mol): 161.16
Molecular Formula: C9H7NO2
InChi Key: ZXLYYQUMYFHCLQ-UHFFFAOYSA-N
SMILES: CN1C(=O)C2=CC=CC=C2C1=O
Percent Purity 
  • (1)
  • (5)
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (4)
  • (1)
  • (5)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)