accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH2OOHFF

CAS RN 55039-89-9

IUPAC Name: 2,2-difluoropent-4-enoic acid
Synonyms: 2,2-difluoropent-4-enoic acid
Molecular Weight (g/mol): 136.1
Molecular Formula: C5H6F2O2
InChi Key: LHOKYUDUAYXFGF-UHFFFAOYSA-N
SMILES: OC(=O)C(F)(F)CC=C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results