CAS RN 5505-72-6
IUPAC Name:
2,4,6-triethenyl-2,4,6-trimethyl-1,3,5,2,4,6-triazatrisilinane
Synonyms:
2,4,6-triethenyl-2,4,6-trimethyl-1,3,5,2,4,6-triazatrisilinane
Molecular Weight (g/mol):
255.54
Molecular Formula:
C9H21N3Si3
InChi Key:
RFSBGZWBVNPVNN-UHFFFAOYSA-N
SMILES:
C[Si]1(N[Si](C)(N[Si](C)(N1)C=C)C=C)C=C
1
–
1
of
1
results
1
2,4,6-Trimethyl-2,4,6-trivinylcyclotrisilazane 97.0+%, TCI America™
CAS: 5505-72-6 Molecular Formula: C9H21N3Si3 Molecular Weight (g/mol): 255.543 MDL Number: MFCD00054934 InChI Key: RFSBGZWBVNPVNN-UHFFFAOYSA-N PubChem CID: 79644 IUPAC Name: 2,4,6-tris(ethenyl)-2,4,6-trimethyl-1,3,5,2,4,6-triazatrisilinane SMILES: C[Si]1(N[Si](N[Si](N1)(C)C=C)(C)C=C)C=C