accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OOCH3

CAS RN 55066-56-3

IUPAC Name: 4-methylphenyl 3-methylbutanoate
Synonyms: P-cresyl isovalerate
Molecular Weight (g/mol): 192.26
Molecular Formula: C12H16O2
InChi Key: MVDPTWHTUYDLTL-UHFFFAOYSA-N
SMILES: CC(C)CC(=O)OC1=CC=C(C)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results