accessibility menu, dialog, popup

UserName

Viewing profile as user:
OH(R)O(S)O(S)OOHO(R)OH(S)OH(S)OH

CAS RN 551-15-5

IUPAC Name: (2S)-7-hydroxy-2-(4-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}phenyl)-3,4-dihydro-2H-1-benzopyran-4-one
Synonyms: liquiritin
Molecular Weight (g/mol): 418.4
Molecular Formula: C21H22O9
InChi Key: DEMKZLAVQYISIA-ZRWXNEIDSA-N
SMILES: OC[C@H]1O[C@@H](OC2=CC=C(C=C2)[C@@H]2CC(=O)C3=CC=C(O)C=C3O2)[C@H](O)[C@@H](O)[C@@H]1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results