accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFF

CAS RN 551-62-2

IUPAC Name: 1,2,3,4-tetrafluorobenzene
Synonyms: 1,2,3,4-tetrafluorobenzene
Molecular Weight (g/mol): 150.08
Molecular Formula: C6H2F4
InChi Key: SOZFIIXUNAKEJP-UHFFFAOYSA-N
SMILES: FC1=CC=C(F)C(F)=C1F
Boiling Point 
  • (2)
  • (3)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (2)
  • (3)
  • (1)
Quantity 
  • (1)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (5)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)