accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 551-93-9

IUPAC Name: 1-(2-aminophenyl)ethan-1-one
Synonyms: O-aminoacetophenone
Molecular Weight (g/mol): 135.17
Molecular Formula: C8H9NO
InChi Key: GTDQGKWDWVUKTI-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=CC=C1N
Boiling Point 
  • (1)
  • (3)
  • (3)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (7)
Percent Purity 
  • (1)
  • (3)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (2)
  • (1)
  • (6)
  • (1)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (2)
  • (2)
  • (7)
  • (1)
  • (3)
  • (5)
  • (4)
  • (1)