accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5511-18-2

H2NO

CAS RN 5511-18-2

IUPAC Name: adamantane-1-carboxamide
Synonyms: adamantane-1-carboxamide
Molecular Weight (g/mol): 179.26
Molecular Formula: C11H17NO
InChi Key: CKBZJTAMRPPVSR-UHFFFAOYSA-N
SMILES: NC(=O)C12CC3CC(CC(C3)C1)C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results