accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 55120-75-7

CaOSOOFFFOSOOFFF

CAS RN 55120-75-7

IUPAC Name: calcium ditrifluoromethanesulfonate
Synonyms: calcium ditriflate
Molecular Weight (g/mol): 338.2
Molecular Formula: C2CaF6O6S2
InChi Key: PUQLFUHLKNBKQQ-UHFFFAOYSA-L
SMILES: [Ca++].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results