accessibility menu, dialog, popup

UserName

Viewing profile as user:
PH2H2P

CAS RN 5518-62-7

IUPAC Name: (2-phosphanylethyl)phosphane
Synonyms: (2-phosphanylethyl)phosphane
Molecular Weight (g/mol): 94.03
Molecular Formula: C2H8P2
InChi Key: FWFZRPMNAAFGBA-UHFFFAOYSA-N
SMILES: PCCP

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results