accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 552-32-9

IUPAC Name: N-(2-nitrophenyl)acetamide
Synonyms: 2-nitroacetanilide
Molecular Weight (g/mol): 180.16
Molecular Formula: C8H8N2O3
InChi Key: BUNFNRVLMKHKIT-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=CC=C1[N+]([O-])=O
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)