accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOHO(S)OOHOH

CAS RN 552-58-9

IUPAC Name: (2S)-2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-3,4-dihydro-2H-1-benzopyran-4-one
Synonyms: eriodictyol eriodictiol
Molecular Weight (g/mol): 288.26
Molecular Formula: C15H12O6
InChi Key: SBHXYTNGIZCORC-ZDUSSCGKSA-N
SMILES: OC1=CC(O)=C2C(=O)C[C@H](OC2=C1)C1=CC=C(O)C(O)=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results